Product Details

SNP ID
rs3729771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19146829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTCGGCAGGTACAGTGGGGGTGG[C/T]GTCTTGCTGGTGAGGTGGCTTGGTG
Phenotype
MIM: 600661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BORCS8-MEF2B PubMed Links

Gene Details

Gene
BORCS8-MEF2B
Gene Name
BORCS8-MEF2B readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005919.3 1315 Silent Mutation ACA,ACG T196T NP_005910.1
Gene
MEF2B
Gene Name
myocyte enhancer factor 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145785.1 1315 Silent Mutation ACA,ACG T196T NP_001139257.1
Gene
TMEM161A
Gene Name
transmembrane protein 161A
There are no transcripts associated with this gene.

View Full Product Details