Product Details
- SNP ID
-
rs3731867
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:218608011 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGATCCCACAGCATCGCAGAGCTC[A/G]GGAGGCACAGCTCACAGACACAGGA
- Phenotype
-
MIM: 605939
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PLCD4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76608525] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PLCD4
- Gene Name
- phospholipase C delta 4
View Full Product Details