Product Details

SNP ID
rs3732337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:30892106 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCAGACGGTATGTGCTGCCTAAT[C/T]AGGACACAGCATTTGCAGTTGTTTT
Phenotype
MIM: 608225
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALNT14 PubMed Links

Gene Details

Gene
GALNT14
Gene Name
polypeptide N-acetylgalactosaminyltransferase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253826.1 Intron NP_001240755.1
NM_001253827.1 Intron NP_001240756.1
NM_024572.3 Intron NP_078848.2
XM_011533105.1 Intron XP_011531407.1
XM_017004906.1 Intron XP_016860395.1
XM_017004907.1 Intron XP_016860396.1
XM_017004908.1 Intron XP_016860397.1

View Full Product Details