Product Details

SNP ID
rs3733403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186265981 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCTAGCAGGCACACAGGCAAAAT[C/G]AAGTTCTACATCTGTCCCTGTGTAT
Phenotype
MIM: 264900 MIM: 229000
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
F11 PubMed Links

Gene Details

Gene
F11
Gene Name
coagulation factor XI
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000128.3 37 UTR 5 NP_000119.1
XM_005262821.3 37 UTR 5 XP_005262878.1
XM_005262822.3 37 UTR 5 XP_005262879.1
XM_005262823.3 37 UTR 5 XP_005262880.1
XM_006714137.2 37 UTR 5 XP_006714200.1
XM_017007884.1 37 UTR 5 XP_016863373.1
XM_017007885.1 37 UTR 5 XP_016863374.1
XM_017007886.1 37 UTR 5 XP_016863375.1
Gene
KLKB1
Gene Name
kallikrein B1
There are no transcripts associated with this gene.

View Full Product Details