Product Details

SNP ID
rs3734616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39190738 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGTCCCGGGCATACATCTAGCTG[A/G]GGATGATGGAAGGTTAGGAAGGCCC
Phenotype
MIM: 603493
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNK5 PubMed Links

Gene Details

Gene
KCNK5
Gene Name
potassium two pore domain channel subfamily K member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003740.3 2016 UTR 3 NP_003731.1
XM_005249456.1 2016 UTR 3 XP_005249513.1
XM_006715235.2 2016 UTR 3 XP_006715298.1

View Full Product Details