Product Details
- SNP ID
-
rs3750731
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:652705 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCAAGCCTCCCATCCCACTTCCC[A/G]GTGCTGACCCCATGAGAACACTTTG
- Phenotype
-
MIM: 611380
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DIP2C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78277958] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DIP2C
- Gene Name
- disco interacting protein 2 homolog C
- Gene
- PRR26
- Gene Name
- proline rich 26
There are no transcripts associated with this gene.
View Full Product Details