Product Details
- SNP ID
-
rs3740855
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:95089940 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCAGAGGCCGCCATGGGCACCGCG[C/T]GCTGGCTCGCGCTGGGCAGCCTCTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ENDOD1
PubMed Links
Gene Details
- Gene
- ENDOD1
- Gene Name
- endonuclease domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015036.2 |
131 |
Missense Mutation |
CGC,TGC |
R5C |
NP_055851.1 |
View Full Product Details