Product Details

SNP ID
rs3743034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40808947 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGCTTGAATAAGGTACTGGAAAC[C/T]GAGGAATTGCTTTACTCACCATCCC
Phenotype
MIM: 616844 MIM: 613256
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC17 PubMed Links

Gene Details

Gene
DNAJC17
Gene Name
DnaJ heat shock protein family (Hsp40) member C17
There are no transcripts associated with this gene.

Gene
PPP1R14D
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14D
There are no transcripts associated with this gene.

Gene
ZFYVE19
Gene Name
zinc finger FYVE-type containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077268.1 1267 Intron NP_001070736.1
NM_001258420.1 1267 Intron NP_001245349.1
NM_001258421.1 1267 Intron NP_001245350.1
NM_032850.4 1267 Intron NP_116239.3
XM_017022683.1 1267 UTR 5 XP_016878172.1
XM_017022684.1 1267 Intron XP_016878173.1

View Full Product Details