Product Details
- SNP ID
-
rs3743219
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:44427835 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCAGTCCTCCTCCTCTTCCAGGG[C/T]CGTCTGCCGGGATCTCCTCCCCTTC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CTDSPL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116908763,rs74658224] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CTDSPL2
- Gene Name
- CTD small phosphatase like 2
View Full Product Details