Product Details

SNP ID
rs3745207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40451456 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGTCATCAGTCACAGCCTGCAGT[C/T]GTGGGGGCACCTTGGTAGGGTCCCA
Phenotype
MIM: 600941 MIM: 612125
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BLVRB PubMed Links

Gene Details

Gene
BLVRB
Gene Name
biliverdin reductase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000713.2 501 Missense Mutation CAA,CGA Q124R NP_000704.1
Gene
LOC105372489
Gene Name
uncharacterized LOC105372489
There are no transcripts associated with this gene.

Gene
SERTAD3
Gene Name
SERTA domain containing 3
There are no transcripts associated with this gene.

View Full Product Details