Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001193268.1 | 2824 | UTR 3 | NP_001180197.1 | ||
NM_001193269.1 | 2824 | UTR 3 | NP_001180198.1 | ||
NM_012155.2 | 2824 | UTR 3 | NP_036287.1 | ||
XM_006723125.2 | 2824 | UTR 3 | XP_006723188.1 | ||
XM_006723126.2 | 2824 | UTR 3 | XP_006723189.1 | ||
XM_011526694.2 | 2824 | UTR 3 | XP_011524996.1 | ||
XM_011526695.2 | 2824 | UTR 3 | XP_011524997.1 | ||
XM_011526698.2 | 2824 | UTR 3 | XP_011525000.1 | ||
XM_011526700.2 | 2824 | UTR 3 | XP_011525002.1 | ||
XM_011526701.2 | 2824 | UTR 3 | XP_011525003.1 | ||
XM_017026550.1 | 2824 | UTR 3 | XP_016882039.1 | ||
XM_017026551.1 | 2824 | Intron | XP_016882040.1 |