Product Details

SNP ID
rs3747888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133281472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGGGGTGCGTGAGCCATGCCCA[C/T]CCCCACCGTGGGCCTTCTTGGCTCC
Phenotype
MIM: 602267
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM8 PubMed Links

Gene Details

Gene
ADAM8
Gene Name
ADAM metallopeptidase domain 8
There are no transcripts associated with this gene.

Gene
TUBGCP2
Gene Name
tubulin gamma complex associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256617.1 Intron NP_001243546.1
NM_001256618.1 Intron NP_001243547.1
NM_006659.3 Intron NP_006650.1
XM_017015595.1 Intron XP_016871084.1
XM_017015596.1 Intron XP_016871085.1

View Full Product Details