Product Details

SNP ID
rs3749977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29112567 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCACTTCTTATGGTGCCATCGTCC[A/G]AGCTATACTGAGGATGCAGTCAACC
Phenotype
MIM: 615016
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2J3 PubMed Links

Gene Details

Gene
OR2J3
Gene Name
olfactory receptor family 2 subfamily J member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005216.3 758 Missense Mutation CAA,CGA Q226R NP_001005216.2

View Full Product Details