Product Details

SNP ID
rs3752220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14566231 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCTTCTTCCGCTGGAGCAGCCTCC[C/G]CTCCCGCTCAAACTCCTTCATGCGC
Phenotype
MIM: 603842 MIM: 610057
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NDUFB7 PubMed Links

Gene Details

Gene
NDUFB7
Gene Name
NADH:ubiquinone oxidoreductase subunit B7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004146.5 393 Missense Mutation CGG,GGG R106G NP_004137.2
XM_011528039.2 393 Missense Mutation CGG,GGG R68G XP_011526341.1
Gene
TECR
Gene Name
trans-2,3-enoyl-CoA reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321170.1 393 Intron NP_001308099.1
NM_138501.5 393 Intron NP_612510.1
XM_006722945.1 393 Intron XP_006723008.1
XM_011528442.2 393 Intron XP_011526744.1

View Full Product Details