Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_019112.3 | Intron | NP_061985.2 | |||
XM_006722616.1 | Intron | XP_006722679.1 | |||
XM_006722617.2 | Intron | XP_006722680.1 | |||
XM_006722618.2 | Intron | XP_006722681.1 | |||
XM_011527628.2 | Intron | XP_011525930.1 | |||
XM_011527629.1 | Intron | XP_011525931.1 | |||
XM_011527630.1 | Intron | XP_011525932.1 | |||
XM_011527631.1 | Intron | XP_011525933.1 | |||
XM_011527632.1 | Intron | XP_011525934.1 | |||
XM_011527633.2 | Intron | XP_011525935.1 | |||
XM_011527634.1 | Intron | XP_011525936.1 | |||
XM_011527635.1 | Intron | XP_011525937.1 | |||
XM_011527636.2 | Intron | XP_011525938.1 | |||
XM_017026142.1 | Intron | XP_016881631.1 | |||
XM_017026143.1 | Intron | XP_016881632.1 |