Product Details

SNP ID
rs3752488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:670093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAGCCATGTGCCCGCGGGCAGC[C/T]TCACTTCACAGCCAGGCTTTGCTTT
Phenotype
MIM: 603264 MIM: 613889
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105371184 PubMed Links

Gene Details

Gene
LOC105371184
Gene Name
uncharacterized LOC105371184
There are no transcripts associated with this gene.

Gene
RHBDL1
Gene Name
rhomboid like 1
There are no transcripts associated with this gene.

Gene
RHOT2
Gene Name
ras homolog family member T2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138769.2 Intron NP_620124.1
XM_005255660.2 Intron XP_005255717.1
XM_005255661.2 Intron XP_005255718.1
XM_005255662.2 Intron XP_005255719.1
XM_005255663.2 Intron XP_005255720.1
XM_006720970.2 Intron XP_006721033.1
XM_006720973.2 Intron XP_006721036.1
XM_017023825.1 Intron XP_016879314.1
XM_017023826.1 Intron XP_016879315.1
XM_017023827.1 Intron XP_016879316.1
XM_017023828.1 Intron XP_016879317.1
XM_017023829.1 Intron XP_016879318.1
XM_017023830.1 Intron XP_016879319.1
XM_017023831.1 Intron XP_016879320.1
XM_017023832.1 Intron XP_016879321.1
XM_017023833.1 Intron XP_016879322.1
XM_017023834.1 Intron XP_016879323.1
XM_017023835.1 Intron XP_016879324.1
XM_017023836.1 Intron XP_016879325.1
XM_017023837.1 Intron XP_016879326.1
XM_017023838.1 Intron XP_016879327.1
XM_017023839.1 Intron XP_016879328.1
XM_017023840.1 Intron XP_016879329.1
XM_017023841.1 Intron XP_016879330.1
XM_017023842.1 Intron XP_016879331.1
Gene
WDR90
Gene Name
WD repeat domain 90
There are no transcripts associated with this gene.

View Full Product Details