Product Details
- SNP ID
-
rs3765604
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:30008247 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCTGAACATGAGGCATAACGAGGT[C/G]CTGGGTTCTGGGCTTCTACCCTGCG
- Phenotype
-
MIM: 615801
MIM: 615714
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HCG8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116244075] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HCG8
- Gene Name
- HLA complex group 8
There are no transcripts associated with this gene.
- Gene
- HLA-J
- Gene Name
- major histocompatibility complex, class I, J (pseudogene)
There are no transcripts associated with this gene.
- Gene
- ZNRD1ASP
- Gene Name
- zinc ribbon domain containing 1 antisense, pseudogene
There are no transcripts associated with this gene.
View Full Product Details