Product Details
- SNP ID
-
rs3761349
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:33589660 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATTTGCACAAGAATTTTTCAAACA[C/T]TTTTCAAATGTGTCAACTGAGCATC
- Phenotype
-
MIM: 603920
MIM: 611428
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CRYZL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1046312] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRYZL1
- Gene Name
- crystallin zeta like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145858.2 |
1392 |
UTR 3 |
|
|
NP_665857.2 |
- Gene
- DONSON
- Gene Name
- downstream neighbor of SON
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017613.3 |
1392 |
Intron |
|
|
NP_060083.1 |
View Full Product Details