Product Details

SNP ID
rs3771889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:151358124 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTGTTTTTATGTCAATGCATTT[A/G]CAATCAGATTTCAGAATCCTAACAG
Phenotype
MIM: 600410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929319 PubMed Links

Gene Details

Gene
LOC101929319
Gene Name
uncharacterized LOC101929319
There are no transcripts associated with this gene.

Gene
TNFAIP6
Gene Name
TNF alpha induced protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007115.3 Intron NP_009046.2

View Full Product Details