Product Details
- SNP ID
-
rs3774394
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:42258051 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATTGTTTTCTTATTCTGCCTCCT[C/G]TGGGTTGGGAGGTTGCTTCCCGTTG
- Phenotype
-
MIM: 118440
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCK
PubMed Links
Gene Details
- Gene
- CCK
- Gene Name
- cholecystokinin
View Full Product Details