Product Details

SNP ID
rs3766353
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:78492174 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGCATTTAACATCAATGAACTTGCT[G/T]GTCCCCTTCCAAAGTTTGGAGATAG
Phenotype
MIM: 600563
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PTGFR PubMed Links

Gene Details

Gene
PTGFR
Gene Name
prostaglandin F receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000959.3 Intron NP_000950.1
NM_001039585.1 Intron NP_001034674.1
XM_017001873.1 Intron XP_016857362.1

View Full Product Details