Product Details

SNP ID
rs3767235
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:20484691 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
TGGAGCTCCTCTCGGAGCCTCTCTG[C/T]GTCTCCGCCTCTCTGGGGCTTTTCC
Phenotype
MIM: 614986
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAMK2N1 PubMed Links

Gene Details

Gene
CAMK2N1
Gene Name
calcium/calmodulin dependent protein kinase II inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018584.5 Intron NP_061054.2

View Full Product Details