Product Details
- SNP ID
-
rs3771548
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:241357313 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACCCTGAGGTGGCAGTGGGTTAGC[A/G]TGTTGGTCTTGTGTAGACTGCCTTA
- Phenotype
-
MIM: 601506
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FARP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10557084] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FARP2
- Gene Name
- FERM, ARH/RhoGEF and pleckstrin domain protein 2
- Gene
- SEPT2
- Gene Name
- septin 2
There are no transcripts associated with this gene.
View Full Product Details