Product Details

SNP ID
rs3788770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:17750922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCATCACACACTAATTTTAGGGA[C/G]TCTAACACTGTGGGTGAGTAAAAGG
Phenotype
MIM: 300227
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SCML1 PubMed Links

Gene Details

Gene
SCML1
Gene Name
sex comb on midleg-like 1 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037535.2 Intron NP_001032624.1
NM_001037536.2 Intron NP_001032625.1
NM_001037540.2 Intron NP_001032629.1
NM_006746.5 Intron NP_006737.2
XM_005274578.2 Intron XP_005274635.1
XM_005274579.2 Intron XP_005274636.1
XM_006724508.3 Intron XP_006724571.1
XM_006724509.3 Intron XP_006724572.1
XM_011545564.2 Intron XP_011543866.1
XM_017029723.1 Intron XP_016885212.1

View Full Product Details