Product Details

SNP ID
rs3776444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:6600942 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCAGGCTCCACCTGTCCAGCCAGG[A/T]GGCATCGACGCCCCAGCAGAGATGG
Phenotype
MIM: 610916
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NSUN2 PubMed Links

Gene Details

Gene
NSUN2
Gene Name
NOP2/Sun RNA methyltransferase family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193455.1 Intron NP_001180384.1
NM_017755.5 Intron NP_060225.4

View Full Product Details