Product Details

SNP ID
rs3210095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27435646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGCAGACCAAGGAGCGAGCTTT[C/T]TCTCAGAGCATGCTTTTCTTTATTA
Phenotype
MIM: 600860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTF3A PubMed Links

Gene Details

Gene
GTF3A
Gene Name
general transcription factor IIIA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002097.2 1754 UTR 3 NP_002088.2
Gene
MTIF3
Gene Name
mitochondrial translational initiation factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166261.1 1754 Intron NP_001159733.1
NM_001166262.1 1754 Intron NP_001159734.1
NM_001166263.1 1754 UTR 3 NP_001159735.1
NM_152912.4 1754 Intron NP_690876.3
XM_006719771.3 1754 UTR 3 XP_006719834.1
XM_006719772.3 1754 UTR 3 XP_006719835.1
XM_011534957.2 1754 UTR 3 XP_011533259.1
XM_011534958.2 1754 UTR 3 XP_011533260.1
XM_011534959.2 1754 UTR 3 XP_011533261.1
XM_011534960.2 1754 UTR 3 XP_011533262.1
XM_011534961.2 1754 UTR 3 XP_011533263.1
XM_011534962.2 1754 UTR 3 XP_011533264.1
XM_011534963.2 1754 UTR 3 XP_011533265.1
XM_017020417.1 1754 Intron XP_016875906.1

View Full Product Details