Product Details

SNP ID
rs3785909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50766609 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCACCAGATTGTAAGAAGTCAATG[A/G]GAAAGTACACAGAAGGTACAACTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00483 PubMed Links

Gene Details

Gene
LINC00483
Gene Name
long intergenic non-protein coding RNA 483
There are no transcripts associated with this gene.

Gene
MIR8059
Gene Name
microRNA 8059
There are no transcripts associated with this gene.

View Full Product Details