Product Details

SNP ID
rs3200457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82614716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAATTAGCTGATTACATGACAG[G/T]GACAGTAAAGAACTAAAGAAACCGG
Phenotype
MIM: 609226
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WDR45B PubMed Links

Gene Details

Gene
WDR45B
Gene Name
WD repeat domain 45B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019613.3 2433 UTR 3 NP_062559.2
XM_005256377.4 2433 Intron XP_005256434.1
XM_017024862.1 2433 Intron XP_016880351.1
XM_017024863.1 2433 Intron XP_016880352.1

View Full Product Details