Product Details

SNP ID
rs3802949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61518481 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTACTTCCTAGAAACGGGGCCCA[A/G]TTGAGTCCTGGGACCCCTCCCTGGC
Phenotype
MIM: 604146
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC10B PubMed Links

Gene Details

Gene
LRRC10B
Gene Name
leucine rich repeat containing 10B
There are no transcripts associated with this gene.

Gene
MIR4488
Gene Name
microRNA 4488
There are no transcripts associated with this gene.

Gene
SYT7
Gene Name
synaptotagmin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252065.1 2263 UTR 3 NP_001238994.1
NM_001300773.1 2263 UTR 3 NP_001287702.1
NM_004200.3 2263 UTR 3 NP_004191.2
XM_005274383.4 2263 UTR 3 XP_005274440.1
XM_005274384.2 2263 UTR 3 XP_005274441.1
XM_005274385.4 2263 UTR 3 XP_005274442.1
XM_005274387.4 2263 UTR 3 XP_005274444.1
XM_005274390.4 2263 UTR 3 XP_005274447.1
XM_006718736.3 2263 UTR 3 XP_006718799.1
XM_011545335.2 2263 UTR 3 XP_011543637.1
XM_011545336.2 2263 UTR 3 XP_011543638.1
XM_011545337.2 2263 UTR 3 XP_011543639.1
XM_011545338.2 2263 UTR 3 XP_011543640.1
XM_011545339.2 2263 UTR 3 XP_011543641.1
XM_011545340.2 2263 UTR 3 XP_011543642.1
XM_011545341.2 2263 UTR 3 XP_011543643.1
XM_011545342.2 2263 Intron XP_011543644.1
XM_011545343.2 2263 UTR 3 XP_011543645.1

View Full Product Details