Product Details
- SNP ID
-
rs3805317
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:140389072 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTTCAAAGTTGCTTCTTTTTCCTC[G/T]GAAATGAAGGACTAAAATAAATGTC
- Phenotype
-
MIM: 601858
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLGN
PubMed Links
Gene Details
- Gene
- CLGN
- Gene Name
- calmegin
- Gene
- SCOC
- Gene Name
- short coiled-coil protein
There are no transcripts associated with this gene.
View Full Product Details