Product Details

SNP ID
rs3807070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:64979744 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAATTTATAAATGAGTTACGTG[C/T]GTGCAGTGTTCCAGGTTATCACAAT
Phenotype
MIM: 194624
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF117 PubMed Links

Gene Details

Gene
ZNF117
Gene Name
zinc finger protein 117
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015852.3 Intron NP_056936.2

View Full Product Details