Product Details

SNP ID
rs3808051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103473615 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATATGTATGTATGTTTTCCCCCAC[A/G]TATGTACATACACACATTTATACAT
Phenotype
MIM: 600514
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927870 PubMed Links

Gene Details

Gene
LOC101927870
Gene Name
uncharacterized LOC101927870
There are no transcripts associated with this gene.

Gene
RELN
Gene Name
reelin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005045.3 Intron NP_005036.2
NM_173054.2 Intron NP_774959.1

View Full Product Details