Product Details

SNP ID
rs3809231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:32107006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAAGCAGAATCCGGAGCCCCTC[C/G]CTACCCGCGGCCGCCGCAGCCCGGG
Phenotype
MIM: 602204
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BICD1 PubMed Links

Gene Details

Gene
BICD1
Gene Name
BICD cargo adaptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003398.1 271 Intron NP_001003398.1
NM_001714.2 271 Intron NP_001705.2
XM_005253465.3 271 UTR 5 XP_005253522.1
XM_005253466.2 271 UTR 5 XP_005253523.1
XM_006719134.3 271 UTR 5 XP_006719197.1
XM_011520812.2 271 UTR 5 XP_011519114.1
XM_011520813.1 271 UTR 5 XP_011519115.1
XM_011520814.2 271 UTR 5 XP_011519116.1
XM_011520815.2 271 UTR 5 XP_011519117.1
XM_011520816.2 271 UTR 5 XP_011519118.1
XM_017019797.1 271 UTR 5 XP_016875286.1

View Full Product Details