Product Details

SNP ID
rs3810315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11887941 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTACGCTATGCCCTGCTGTAGTCA[C/T]AGGAGGTGTAGAGAGGACCCCGGGA
Phenotype
MIM: 194543
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF69 PubMed Links

Gene Details

Gene
ZNF69
Gene Name
zinc finger protein 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321163.1 169 Silent Mutation CAC,CAT H6H NP_001308092.1
NM_021915.3 169 Silent Mutation CAC,CAT H6H NP_068734.1
XM_006722873.3 169 Intron XP_006722936.1
XM_006722874.3 169 Silent Mutation CAC,CAT H6H XP_006722937.1
XM_017027231.1 169 Silent Mutation CAC,CAT H6H XP_016882720.1

View Full Product Details