Product Details

SNP ID
rs3811025
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:157796683 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGAATTATTCAAAGTTTACCAAATG[C/T]TAATTTTTAGTGACACAAATTACTA
Phenotype
MIM: 606508
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCRL1 PubMed Links

Gene Details

Gene
FCRL1
Gene Name
Fc receptor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159397.1 Intron NP_001152869.1
NM_001159398.1 Intron NP_001152870.1
NM_052938.4 Intron NP_443170.1
XM_005244866.2 Intron XP_005244923.1
XM_005244867.2 Intron XP_005244924.1
XM_005244869.3 Intron XP_005244926.1
XM_011509125.1 Intron XP_011507427.1
XM_011509126.1 Intron XP_011507428.1
XM_011509127.2 Intron XP_011507429.1
XM_011509128.1 Intron XP_011507430.1
XM_011509129.1 Intron XP_011507431.1
XM_011509130.1 Intron XP_011507432.1
XM_011509131.1 Intron XP_011507433.1
XM_011509132.2 Intron XP_011507434.1
XM_011509133.2 Intron XP_011507435.1
XM_011509134.2 Intron XP_011507436.1
XM_011509135.2 Intron XP_011507437.1
XM_011509136.1 Intron XP_011507438.1
XM_011509137.2 Intron XP_011507439.1
XM_017000227.1 Intron XP_016855716.1

View Full Product Details