Product Details

SNP ID
rs3813315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:137619172 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATCTGGCTCAGAAATATACAAAC[A/G]TTACACAACAGATATCAAAGTAAGA
Phenotype
MIM: 605775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL3 PubMed Links

Gene Details

Gene
KLHL3
Gene Name
kelch like family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257194.1 5134 UTR 3 NP_001244123.1
NM_001257195.1 5134 UTR 3 NP_001244124.1
NM_017415.2 5134 UTR 3 NP_059111.2

View Full Product Details