Product Details

SNP ID
rs3795509
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:20554816 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TACCTAGAAAGTAGACGCAGCAGAG[A/T]TGCTCCCAAGTCGAGGCTCCTCAGA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM43B PubMed Links

Gene Details

Gene
FAM43B
Gene Name
family with sequence similarity 43 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207334.2 2378 UTR 3 NP_997217.1

View Full Product Details