Product Details

SNP ID
rs3735320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149549589 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGACAAACACTGCCACTTTCTAAGG[A/T]GCAAGTCAATTTCTTTTCAAGTTTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF767P PubMed Links

Gene Details

Gene
ZNF767P
Gene Name
zinc finger family member 767, pseudogene
There are no transcripts associated with this gene.

View Full Product Details