Product Details

SNP ID
rs3801393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:42922879 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCATTAGAGCAGACAACTTACC[C/T]AAGGTACCAATTACACAATCATTTT
Phenotype
MIM: 611839 MIM: 176842
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf25 PubMed Links

Gene Details

Gene
C7orf25
Gene Name
chromosome 7 open reading frame 25
There are no transcripts associated with this gene.

Gene
MRPL32
Gene Name
mitochondrial ribosomal protein L32
There are no transcripts associated with this gene.

Gene
PSMA2
Gene Name
proteasome subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002787.4 Intron NP_002778.1

View Full Product Details