Product Details

SNP ID
rs3738212
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:167431062 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGGTGCGGCACAGAGGCCTGAGGCA[C/G]GGAGGCTCCCGCTGCCCTCGCAGGC
Phenotype
MIM: 186780 MIM: 164175
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD247 PubMed Links

Gene Details

Gene
CD247
Gene Name
CD247 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000734.3 1333 UTR 3 NP_000725.1
NM_198053.2 1333 UTR 3 NP_932170.1
XM_011510144.2 1333 UTR 3 XP_011508446.1
XM_011510145.1 1333 UTR 3 XP_011508447.1
XM_017002800.1 1333 UTR 3 XP_016858289.1
XM_017002801.1 1333 UTR 3 XP_016858290.1
Gene
POU2F1
Gene Name
POU class 2 homeobox 1
There are no transcripts associated with this gene.

View Full Product Details