Product Details

SNP ID
rs3802611
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:26748839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACAATTAAATATATAGCACAGC[A/T]AAACTATTTTTATGAAGATTTCTTA
Phenotype
MIM: 603050 MIM: 607429
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
ABI1 PubMed Links

Gene Details

Gene
ABI1
Gene Name
abl interactor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012750.2 Intron NP_001012768.1
NM_001012751.2 Intron NP_001012769.1
NM_001012752.2 Intron NP_001012770.1
NM_001178116.1 Intron NP_001171587.1
NM_001178119.1 Intron NP_001171590.1
NM_001178120.1 Intron NP_001171591.1
NM_001178121.1 Intron NP_001171592.1
NM_001178122.1 Intron NP_001171593.1
NM_001178123.1 Intron NP_001171594.1
NM_001178124.1 Intron NP_001171595.1
NM_001178125.1 Intron NP_001171596.1
NM_005470.3 Intron NP_005461.2
XM_005252332.3 Intron XP_005252389.1
XM_005252333.3 Intron XP_005252390.1
XM_006717359.2 Intron XP_006717422.1
XM_006717360.2 Intron XP_006717423.1
XM_006717361.2 Intron XP_006717424.1
XM_011519289.2 Intron XP_011517591.1
XM_017015458.1 Intron XP_016870947.1
XM_017015459.1 Intron XP_016870948.1
Gene
PDSS1
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 1
There are no transcripts associated with this gene.

View Full Product Details