Product Details

SNP ID
rs3738958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223876005 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCAATAGTATTTCTTTATATCCT[A/T]TAGGTAGCAGGACAGAGCTAGGACA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
WDFY1 PubMed Links

Gene Details

Gene
WDFY1
Gene Name
WD repeat and FYVE domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020830.4 4002 UTR 3 NP_065881.1

View Full Product Details