Product Details

SNP ID
rs3803685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89953948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCAACATTGGGCAAGAGGACCC[C/T]GTGCTGGGAGGCAGGGAGCTTGGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEF8 PubMed Links

Gene Details

Gene
DEF8
Gene Name
differentially expressed in FDCP 8 homolog (mouse)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242816.1 Intron NP_001229745.1
NM_001242817.1 Intron NP_001229746.1
NM_001242818.1 Intron NP_001229747.1
NM_001242819.1 Intron NP_001229748.1
NM_001242820.1 Intron NP_001229749.1
NM_001242821.1 Intron NP_001229750.1
NM_001242822.1 Intron NP_001229751.1
NM_017702.3 Intron NP_060172.1
NM_207514.2 Intron NP_997397.1
XM_005256318.2 Intron XP_005256375.2
XM_011523156.2 Intron XP_011521458.1
XM_011523157.2 Intron XP_011521459.1
XM_011523158.1 Intron XP_011521460.1
XM_011523159.2 Intron XP_011521461.1
XM_011523160.2 Intron XP_011521462.1
XM_011523161.2 Intron XP_011521463.1
XM_011523162.2 Intron XP_011521464.1
XM_017023358.1 Intron XP_016878847.1
XM_017023359.1 Intron XP_016878848.1
XM_017023360.1 Intron XP_016878849.1
XM_017023361.1 Intron XP_016878850.1
XM_017023362.1 Intron XP_016878851.1
XM_017023363.1 Intron XP_016878852.1
XM_017023364.1 Intron XP_016878853.1
XM_017023365.1 Intron XP_016878854.1
XM_017023366.1 Intron XP_016878855.1
XM_017023367.1 Intron XP_016878856.1
XM_017023368.1 Intron XP_016878857.1
XM_017023369.1 Intron XP_016878858.1

View Full Product Details