Product Details

SNP ID
rs3806755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:985242 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGTGTGAGGAACGAACGCAGCAG[A/C]GTGTCGCCATGTCTTGTGTGGCTGG
Phenotype
MIM: 252800 MIM: 610130
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IDUA PubMed Links

Gene Details

Gene
IDUA
Gene Name
iduronidase, alpha-L-
There are no transcripts associated with this gene.

Gene
SLC26A1
Gene Name
solute carrier family 26 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022042.3 Intron NP_071325.2
NM_134425.2 Intron NP_602297.1
NM_213613.3 Intron NP_998778.1

View Full Product Details