Product Details

SNP ID
rs3740751
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:101030868 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCACATAGCAAGAGGAAGTGAGAG[A/G]CTTTCATGATTCAGAAGAAGGAAGA
Phenotype
MIM: 607311
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PGR PubMed Links

Gene Details

Gene
PGR
Gene Name
progesterone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000926.4 11793 UTR 3 NP_000917.3
NM_001202474.3 11793 UTR 3 NP_001189403.1
NM_001271161.2 11793 UTR 3 NP_001258090.1
NM_001271162.1 11793 UTR 3 NP_001258091.1
XM_006718858.3 11793 Intron XP_006718921.1
XM_011542869.2 11793 Intron XP_011541171.1

View Full Product Details