Product Details

SNP ID
rs3810219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5690405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGTGCAGGTTGGAGGATGGTTG[A/G]TTGTGGCGAGCGAGGCTGAAGGAGC
Phenotype
MIM: 616658 MIM: 605490
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf70 PubMed Links

Gene Details

Gene
C19orf70
Gene Name
chromosome 19 open reading frame 70
There are no transcripts associated with this gene.

Gene
HSD11B1L
Gene Name
hydroxysteroid 11-beta dehydrogenase 1 like
There are no transcripts associated with this gene.

Gene
LONP1
Gene Name
lon peptidase 1, mitochondrial
There are no transcripts associated with this gene.

Gene
RPL36
Gene Name
ribosomal protein L36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015414.3 Intron NP_056229.2
NM_033643.2 Intron NP_378669.1

View Full Product Details