Product Details

SNP ID
rs3743742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68311133 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGGTATGCTGGGAAGGTGGGGTC[G/T]CTTTGGGGTTCTGTGTGCAGCGAGC
Phenotype
MIM: 610087 MIM: 605641
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRMT7 PubMed Links

Gene Details

Gene
PRMT7
Gene Name
protein arginine methyltransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184824.1 20 Intron NP_001171753.1
NM_001290018.1 20 UTR 5 NP_001276947.1
NM_019023.2 20 Intron NP_061896.1
XM_011523112.2 20 Intron XP_011521414.1
XM_011523113.2 20 Intron XP_011521415.1
XM_011523115.2 20 Intron XP_011521417.1
XM_011523116.2 20 Intron XP_011521418.1
XM_011523121.2 20 Intron XP_011521423.1
XM_011523124.2 20 Intron XP_011521426.1
XM_011523125.2 20 Intron XP_011521427.1
XM_011523126.2 20 Intron XP_011521428.1
XM_011523128.2 20 Intron XP_011521430.1
XM_011523131.2 20 Intron XP_011521433.1
XM_017023290.1 20 Intron XP_016878779.1
XM_017023291.1 20 Intron XP_016878780.1
XM_017023292.1 20 Intron XP_016878781.1
XM_017023293.1 20 Intron XP_016878782.1
XM_017023294.1 20 Intron XP_016878783.1
XM_017023295.1 20 Intron XP_016878784.1
XM_017023296.1 20 Intron XP_016878785.1
XM_017023297.1 20 Intron XP_016878786.1
XM_017023298.1 20 Intron XP_016878787.1
XM_017023299.1 20 Intron XP_016878788.1
XM_017023300.1 20 Intron XP_016878789.1
XM_017023301.1 20 Intron XP_016878790.1
XM_017023302.1 20 Intron XP_016878791.1
XM_017023303.1 20 Intron XP_016878792.1
XM_017023304.1 20 Intron XP_016878793.1
XM_017023305.1 20 Intron XP_016878794.1
XM_017023306.1 20 Intron XP_016878795.1
XM_017023307.1 20 Intron XP_016878796.1
XM_017023308.1 20 Intron XP_016878797.1
XM_017023309.1 20 Intron XP_016878798.1
XM_017023310.1 20 Intron XP_016878799.1
XM_017023311.1 20 Intron XP_016878800.1
XM_017023312.1 20 Intron XP_016878801.1
XM_017023313.1 20 Intron XP_016878802.1
XM_017023314.1 20 Intron XP_016878803.1
XM_017023315.1 20 Intron XP_016878804.1
XM_017023316.1 20 Intron XP_016878805.1
Gene
SLC7A6
Gene Name
solute carrier family 7 member 6
There are no transcripts associated with this gene.

Gene
SLC7A6OS
Gene Name
solute carrier family 7 member 6 opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032178.2 20 Intron NP_115554.2
XM_011523372.2 20 Intron XP_011521674.1

View Full Product Details