Product Details

SNP ID
rs3813576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78268067 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCCCATCCCATGTGAGATCTGGC[C/T]TCTGACTGAATGTCCCAAGCAACTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJA4 PubMed Links

Gene Details

Gene
DNAJA4
Gene Name
DnaJ heat shock protein family (Hsp40) member A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130182.1 Intron NP_001123654.1
NM_001130183.1 Intron NP_001123655.1
NM_018602.3 Intron NP_061072.3
XM_005254535.3 Intron XP_005254592.1
XM_006720601.1 Intron XP_006720664.1

View Full Product Details