Product Details

SNP ID
rs3745642
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:29923680 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACTCTTTTCCAGGCTCCTTGGAG[G/T]TCACTGCCCTGATGACAACATGGTC
Phenotype
MIM: 603494
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
URI1 PubMed Links

Gene Details

Gene
URI1
Gene Name
URI1, prefoldin like chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252641.1 31 UTR 5 NP_001239570.1
NM_003796.3 31 Intron NP_003787.2
XM_005259362.2 31 UTR 5 XP_005259419.1
XM_005259363.4 31 Intron XP_005259420.1
XM_011527435.1 31 Intron XP_011525737.1

View Full Product Details