Product Details

SNP ID
rs3748395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87327021 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACGGATTTACTTGAAACTGTGAG[A/G]AGAAACAAGTGCACGGGTGCAGGTG
Phenotype
MIM: 609102
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf95 PubMed Links

Gene Details

Gene
C16orf95
Gene Name
chromosome 16 open reading frame 95
There are no transcripts associated with this gene.

Gene
FBXO31
Gene Name
F-box protein 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282683.1 5936 UTR 3 NP_001269612.1
NM_024735.4 5936 UTR 3 NP_079011.3
Gene
LOC101928659
Gene Name
uncharacterized LOC101928659
There are no transcripts associated with this gene.

View Full Product Details